missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxycatechol 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 125811000

 View more versions of this product

Catalog No. AC125811000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Methoxycatechol
39° to 43°C
99%
98.5
Authentic
Crystalline Solid
99%
140.14
06, 1081
Slightly beige to light brown
100g
934-00-9
100.0
Technical Grade
≥98.5%
C7H8O3
MFCD00002191
Pyrogallol monomethyl ether

Chemical Identifiers

934-00-9
100.0
C7H8O3
MFCD00002191
3-Methoxycatechol, 99%
Technical Grade
98.5
99%
140.14
Pyrogallol monomethyl ether
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-276-4

RTECSNumber : CZ9002750

TSCA : TSCA