Please login to your online account to display your discounted pricing

3-Methoxycatechol 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methoxycatechol
Name Note 99%
CAS 934-00-9
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8O3
Formula Weight 140.14
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC125811000

  SDS
thermo fisher scientific
125811000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.14
Assay ≥98.5%
Purity Grade Technical Grade
Physical Form Crystalline Solid
Quantity 100g
Color Slightly beige to light brown
EINECS Number 213-276-4
RTECS Number CZ9002750
TSCA TSCA
Melting Point 39° to 43°C
Beilstein 06, 1081
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 98.5
Synonym Pyrogallol monomethyl ether
Name Note 99%
Chemical Name or Material 3-Methoxycatechol
Molecular Formula C7H8O3
CAS 934-00-9
Assay Percent Range 99%
MDL Number MFCD00002191
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.