Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methoxycarbonyl)phenyl isothiocyanate, 99%

Chemical Identifiers

Chemical Name or Material 3-(Methoxycarbonyl)phenyl isothiocyanate
CAS 3125-66-4
Molecular Formula C9H7NO2S
Formula Weight 193.23
MDL Number MFCD00041080
UN Number UN2922
Beilstein 2100452
Synonym Methyl 3-isothiocyanatobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1047603

 
alfa aesar™
L1047603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1047606

 
alfa aesar™
L1047606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.23
Chemical Name or Material 3-(Methoxycarbonyl)phenyl isothiocyanate
Assay 99%
Molecular Formula C9H7NO2S
MDL Number MFCD00041080
CAS 3125-66-4
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Odor Irritating
Boiling Point 105° to 106°C (0.45mmHg)
UN Number UN2922
Beilstein 2100452
Synonym Methyl 3-isothiocyanatobenzoate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.