Learn More
3-(Methoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1870903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(Methoxycarbonyl)phenyl isocyanate | |
35°C to 38°C | |
C9H7NO3 | |
1 g | |
Moisture sensitive | |
WBGWGERFPSYHDT-UHFFFAOYSA-N | |
methyl 3-isocyanatobenzoate | |
2733366 | |
97% |
41221-47-0 | |
>110°C (230°F) | |
MFCD00037077 | |
UN2811 | |
3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate | |
COC(=O)C1=CC=CC(=C1)N=C=O | |
177.16 | |
177.16 |
Chemical Identifiers
41221-47-0 | |
177.16 | |
WBGWGERFPSYHDT-UHFFFAOYSA-N | |
2733366 | |
COC(=O)C1=CC=CC(=C1)N=C=O |
C9H7NO3 | |
MFCD00037077 | |
3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate | |
methyl 3-isocyanatobenzoate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only