Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-(Methoxycarbonyl)biphenyl-4-boronic acid pinacol ester, 95%

$236.74 - $236.74

Chemical Identifiers

Chemical Name or Material 3'-(Methoxycarbonyl)biphenyl-4-boronic acid pinacol ester
CAS 1256358-85-6
Molecular Formula C20H23BO4
Formula Weight 338.21g/mol
MDL Number MFCD16294535
Synonym 4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-3-carboxylic acid methyl ester; 4-(3-Methoxycarbonylphenyl)benzeneboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5191606

 
alfa aesar™
H51916-06
Each for $236.74
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 3'-(Methoxycarbonyl)biphenyl-4-boronic acid pinacol ester
CAS 1256358-85-6
Assay 95%
Molecular Formula C20H23BO4
Formula Weight 338.21g/mol
MDL Number MFCD16294535
Synonym 4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-3-carboxylic acid methyl ester; 4-(3-Methoxycarbonylphenyl)benzeneboronic acid pinacol ester