Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxycarbonyl-3-sulfolene, 98%


Quantity 1g

Manufacturer: alfa aesar™  L1835303

 View more versions of this product

Catalog No. AAL1835303

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 176.19
Chemical Name or Material 3-Methoxycarbonyl-3-sulfolene
Beilstein 1368581
Melting Point 60° to 64°C
Assay 98%
Molecular Formula C6H8O4S
MDL Number MFCD00042754
Synonym 3-Carbomethoxy-3-sulfolene; 3-Methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide
Quantity 1g
CAS 67488-50-0

Specifications

Chemical Name or Material 3-Methoxycarbonyl-3-sulfolene
CAS 67488-50-0
Molecular Formula C6H8O4S
Formula Weight 176.19
MDL Number MFCD00042754
Beilstein 1368581
Synonym 3-Carbomethoxy-3-sulfolene; 3-Methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide