Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxycarbonyl-3-sulfolene, 98%

$103.07 - $103.07

Chemical Identifiers

Chemical Name or Material 3-Methoxycarbonyl-3-sulfolene
CAS 67488-50-0
Molecular Formula C6H8O4S
Formula Weight 176.19
MDL Number MFCD00042754
Beilstein 1368581
Synonym 3-Carbomethoxy-3-sulfolene; 3-Methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1835303

  SDS
alfa aesar™
L1835303
1g Each for $103.07
Description & Specifications

Specifications

Formula Weight 176.19
Beilstein 1368581
Melting Point 60° to 64°C
Chemical Name or Material 3-Methoxycarbonyl-3-sulfolene
MDL Number MFCD00042754
Assay 98%
Molecular Formula C6H8O4S
Quantity 1g
Synonym 3-Carbomethoxy-3-sulfolene; 3-Methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide
CAS 67488-50-0