missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methoxycarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5317103

 View more versions of this product

Catalog No. AAH5317103



Description

Description

Applications
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

3-(Methoxycarbamoyl)benzeneboronic acid
850567-26-9
MFCD04115698
NDJZQOMZZXSXEM-UHFFFAOYSA-N
[3-(methoxycarbamoyl)phenyl]boronic acid
44118753
97%
1g
C8H10BNO4
3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, 3-methoxycarbamoyl phenyl boronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid
B(C1=CC(=CC=C1)C(=O)NOC)(O)O
194.981
194.98

Chemical Identifiers

850567-26-9
194.981
NDJZQOMZZXSXEM-UHFFFAOYSA-N
44118753
B(C1=CC(=CC=C1)C(=O)NOC)(O)O
C8H10BNO4
MFCD04115698
3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, 3-methoxycarbamoyl phenyl boronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid
[3-(methoxycarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents