Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Methoxybiphenyl-3-carboxylic acid, 95%

$72.97 - $233.97

Chemical Identifiers

Chemical Name or Material 3'-Methoxybiphenyl-3-carboxylic acid
CAS 168618-45-9
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD03424598
Synonym 3-(3-Methoxyphenyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5176703

 
alfa aesar™
H5176703
1g Each for $72.97

AAH5176706

 
alfa aesar™
H5176706
5g Each for $233.97
Description & Specifications

Specifications

Formula Weight 228.25
Chemical Name or Material 3'-Methoxybiphenyl-3-carboxylic acid
Assay 95%
Molecular Formula C14H12O3
MDL Number MFCD03424598
CAS 168618-45-9
Synonym 3-(3-Methoxyphenyl)benzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.