Learn More
3-Methoxybenzyl alcohol, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1547114
Description
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in ether, and alcohol. Insoluble in water.
Notes
Incompatible with Oxidizing agents, acids, acid chlorides, acid anhydrides.
Specifications
3-Methoxybenzyl alcohol | |
1.112 | |
109°C (228°F) | |
1.544 | |
25 g | |
3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol | |
IIGNZLVHOZEOPV-UHFFFAOYSA-N | |
(3-methoxyphenyl)methanol | |
81437 | |
138.17 |
6971-51-3 | |
252°C to 254°C | |
C8H10O2 | |
MFCD00004637 | |
2042063 | |
Soluble in ether,and alcohol. Insoluble in water. | |
COC1=CC=CC(=C1)CO | |
138.166 | |
CHEBI:1580 | |
≥98% |
Chemical Identifiers
6971-51-3 | |
138.166 | |
IIGNZLVHOZEOPV-UHFFFAOYSA-N | |
81437 | |
(3-methoxyphenyl)methanol |
C8H10O2 | |
MFCD00004637 | |
3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol | |
CHEBI:1580 | |
COC1=CC=CC(=C1)CO |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-200-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only