missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxybenzotrifluoride, TCI America™

$50.42 - $151.25

Chemical Identifiers

CAS 454-90-0
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00000385
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
PubChem CID 96691
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
SMILES COC1=CC=CC(=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22435G View Documents TCI America
M22435G
5g Each for $50.42
 
M224325G View Documents TCI America
M224325G
25g Each for $151.25
 
Specifications

Chemical Identifiers

454-90-0
176.138
XHONYVFDZSPELQ-UHFFFAOYSA-N
96691
COC1=CC=CC(=C1)C(F)(F)F
C8H7F3O
MFCD00000385
3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
1-methoxy-3-(trifluoromethyl)benzene

Specifications

160°C
5g
C8H7F3O
1993
XHONYVFDZSPELQ-UHFFFAOYSA-N
1-methoxy-3-(trifluoromethyl)benzene
96691
Clear Liquid at 20°C
3-Methoxybenzotrifluoride
Yellow
454-90-0
MFCD00000385
3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
COC1=CC=CC(=C1)C(F)(F)F
176.138
176.14
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No