Please login to your online account to display your discounted pricing

3-Methoxybenzonitrile, 98%, Acros Organics

$243.64 - $243.64

Chemical Identifiers

Chemical Name or Material 3-Methoxybenzonitrile
Name Note 98%
CAS 1527-89-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7NO
Linear Formula CH3OC6H4CN
Formula Weight 133.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183730250

  SDS
thermo fisher scientific
183730250
Glass bottle 25mL Each for $243.64
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 133.15
Physical Form Liquid
Quantity 25mL
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 216-201-3
Density 1.089g/mL
Beilstein 10, II, 82
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
CAS Min % 97.5
Refractive Index 1.5392 to 1.5412 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Name Note 98%
Linear Formula CH3OC6H4CN
MDL Number MFCD00001801
Chemical Name or Material 3-Methoxybenzonitrile
Assay Percent Range 98%
Molecular Formula C8H7NO
CAS 1527-89-5