Learn More
3-Methoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1705003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methoxybenzenesulfonyl chloride | |
1.46 | |
>110°C (230°F) | |
1.557 | |
1 g | |
2093871 | |
3-methoxybenzene-1-sulfonyl chloride, 3-methoxy-benzenesulfonyl chloride, 3-methoxybenzenesulphonyl chloride, m-anisolesulfonyl chloride, 3-methoxybenzenesulfonylchloride, benzenesulfonyl chloride, 3-methoxy, akos bb-9459, 3-methoxyphenylsulfonyl chloride, 3-methoxybenzene sulfonyl chloride, m-anisolesulphonyl chloride | |
COC1=CC(=CC=C1)S(=O)(=O)Cl | |
206.64 | |
206.65 |
10130-74-2 | |
158°C to 159°C (20 mmHg) | |
C7H7ClO3S | |
MFCD01318155 | |
UN3265 | |
Moisture sensitive | |
JHJKSEKUZNJKGO-UHFFFAOYSA-N | |
3-methoxybenzenesulfonyl chloride | |
2737504 | |
97% |
Chemical Identifiers
10130-74-2 | |
206.64 | |
JHJKSEKUZNJKGO-UHFFFAOYSA-N | |
2737504 | |
COC1=CC(=CC=C1)S(=O)(=O)Cl |
C7H7ClO3S | |
MFCD01318155 | |
3-methoxybenzene-1-sulfonyl chloride, 3-methoxy-benzenesulfonyl chloride, 3-methoxybenzenesulphonyl chloride, m-anisolesulfonyl chloride, 3-methoxybenzenesulfonylchloride, benzenesulfonyl chloride, 3-methoxy, akos bb-9459, 3-methoxyphenylsulfonyl chloride, 3-methoxybenzene sulfonyl chloride, m-anisolesulphonyl chloride | |
3-methoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only