Learn More
3-Methoxybenzenesulfonyl chloride, 96%, ACROS Organics™
Chemical Identifiers
CAS | 10130-74-2 |
---|---|
Molecular Formula | C7H7ClO3S |
Molecular Weight (g/mol) | 206.65 |
MDL Number | MFCD01318155 |
InChI Key | JHJKSEKUZNJKGO-UHFFFAOYSA-N |
Synonym | 3-methoxy-benzenesulfonyl chloride, 3-methoxybenzene sulfonyl chloride, 3-methoxybenzene-1-sulfonyl chloride, 3-methoxybenzenesulfonylchloride, 3-methoxybenzenesulphonyl chloride, 3-methoxyphenylsulfonyl chloride, akos bb-9459, benzenesulfonyl chloride, 3-methoxy, m-anisolesulfonyl chloride, m-anisolesulphonyl chloride |
PubChem CID | 2737504 |
IUPAC Name | 3-methoxybenzenesulfonyl chloride |
SMILES | COC1=CC(=CC=C1)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398520010
|
Thermo Fisher Scientific
AC398520010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC398520050
|
Thermo Fisher Scientific
AC398520050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
10130-74-2 | |
206.65 | |
JHJKSEKUZNJKGO-UHFFFAOYSA-N | |
2737504 | |
COC1=CC(=CC=C1)S(=O)(=O)Cl |
C7H7ClO3S | |
MFCD01318155 | |
3-methoxy-benzenesulfonyl chloride, 3-methoxybenzene sulfonyl chloride, 3-methoxybenzene-1-sulfonyl chloride, 3-methoxybenzenesulfonylchloride, 3-methoxybenzenesulphonyl chloride, 3-methoxyphenylsulfonyl chloride, akos bb-9459, benzenesulfonyl chloride, 3-methoxy, m-anisolesulfonyl chloride, m-anisolesulphonyl chloride | |
3-methoxybenzenesulfonyl chloride |
Specifications
Authentic | |
1.5550 to 1.5570 (20°C, 589nm) | |
1g | |
95.0 | |
95% min. (GC) | |
CH3OC6H4SO2Cl | |
3-methoxy-benzenesulfonyl chloride, 3-methoxybenzene sulfonyl chloride, 3-methoxybenzene-1-sulfonyl chloride, 3-methoxybenzenesulfonylchloride, 3-methoxybenzenesulphonyl chloride, 3-methoxyphenylsulfonyl chloride, akos bb-9459, benzenesulfonyl chloride, 3-methoxy, m-anisolesulfonyl chloride, m-anisolesulphonyl chloride | |
COC1=CC(=CC=C1)S(=O)(=O)Cl | |
206.65 | |
206.65 | |
96% |
Glass bottle | |
Brown to Yellow | |
10130-74-2 | |
100.0 | |
C7H7ClO3S | |
MFCD01318155 | |
JHJKSEKUZNJKGO-UHFFFAOYSA-N | |
3-methoxybenzenesulfonyl chloride | |
2737504 | |
Liquid | |
3-Methoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger