Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxybenzamide, 97%

$51.38 - $176.30

Chemical Identifiers

Chemical Name or Material 3-Methoxybenzamide
CAS 5813-86-5
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00007986
Beilstein 2206857
Synonym m-Anisamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2320406

  SDS
alfa aesar™
B2320406
5g Each for $51.38

AAB2320414

  SDS
alfa aesar™
B2320414
25g Each for $176.30
Description & Specifications

Specifications

Formula Weight 151.17
Quantity 5g
Melting Point 131° to 135°C
Molecular Formula C8H9NO2
Beilstein 2206857
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 3-Methoxybenzamide
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 5813-86-5
RTECS Number CV5463333
MDL Number MFCD00007986
Synonym m-Anisamide

Applications
3-Methoxybenzamide is act as an inhibitor of ADP-ribosyltransferase.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.