Learn More
3-Methoxyazetidine hydrochloride, 95%, Thermo Scientific Chemicals
$98.81 - $194.40
Chemical Identifiers
CAS | 148644-09-1 |
---|---|
Molecular Formula | C4H10ClNO |
Molecular Weight (g/mol) | 123.58 |
MDL Number | MFCD06804514 |
InChI Key | KSXGQRBTBLQJEF-UHFFFAOYSA-N |
Synonym | 3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride |
PubChem CID | 22242858 |
IUPAC Name | 3-methoxyazetidine;hydrochloride |
SMILES | COC1CNC1.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57769MD
|
Thermo Scientific Chemicals
H57769MD |
250 mg |
Each for $98.81
|
|
|||||
AAH57769ME
|
Thermo Scientific Chemicals
H57769ME |
500 mg |
Each for $194.40
|
|
|||||
Description
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
Chemical Identifiers
148644-09-1 | |
123.58 | |
KSXGQRBTBLQJEF-UHFFFAOYSA-N | |
22242858 | |
COC1CNC1.Cl |
C4H10ClNO | |
MFCD06804514 | |
3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride | |
3-methoxyazetidine;hydrochloride |
Specifications
148644-09-1 | |
C4H10ClNO | |
250 mg | |
KSXGQRBTBLQJEF-UHFFFAOYSA-N | |
3-methoxyazetidine;hydrochloride | |
22242858 | |
95% |
143°C to 149°C | |
MFCD06804514 | |
3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride | |
COC1CNC1.Cl | |
123.58 | |
123.58 | |
3-Methoxyazetidine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only