missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Methoxyacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1560414

 View more versions of this product

Catalog No. AAA1560414



Description

Description

Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

Solubility
Fully miscible in water. Soluble in alcohol.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3'-Methoxyacetophenone
239°C to 241°C
25g
C9H10O2
878552
BAYUSCHCCGXLAY-UHFFFAOYSA-N
1-(3-methoxyphenyl)ethanone
11460
97%
>110°C (230°F)
-7°C
586-37-8
MFCD00008736
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
CC(=O)C1=CC(=CC=C1)OC
150.177
150.18

Chemical Identifiers

586-37-8
150.177
BAYUSCHCCGXLAY-UHFFFAOYSA-N
11460
CC(=O)C1=CC(=CC=C1)OC
C9H10O2
MFCD00008736
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
1-(3-methoxyphenyl)ethanone
Documents