missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-N,N-dimethylbenzylamine, 98%

Manufacturer:  Alfa Aesar™ H5526003

 View more versions of this product

Catalog No. AAH5526003



Specifications

Specifications

3-Methoxy-N,N-dimethylbenzylamine
96°C (205°F)
105°C (13mmHg)
1g
C10H15NO
1-3-methoxyphenyl-n,n-dimethylmethanamine
CN(C)CC1=CC(=CC=C1)OC
165.236
165.24
98%
0.984 g/mL
1.514
Colorless
15184-99-3
MFCD00011530
IENUHHADGQLQLU-UHFFFAOYSA-N
1-(3-methoxyphenyl)-N,N-dimethylmethanamine
547434
Liquid

Chemical Identifiers

15184-99-3
165.236
IENUHHADGQLQLU-UHFFFAOYSA-N
547434
CN(C)CC1=CC(=CC=C1)OC
C10H15NO
MFCD00011530
1-3-methoxyphenyl-n,n-dimethylmethanamine
1-(3-methoxyphenyl)-N,N-dimethylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents