missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-N,N-dimethylbenzylamine, 98%

Manufacturer:  Alfa Aesar™ H5526014

 View more versions of this product

Catalog No. AAH5526014


 

Specifications

Specifications

3-Methoxy-N,N-dimethylbenzylamine
105°C (13mmHg)
25g
1.5140
C10H15NO
IENUHHADGQLQLU-UHFFFAOYSA-N
1-(3-methoxyphenyl)-N,N-dimethylmethanamine
547434
Liquid
0.984g/mL
Colorless
15184-99-3
98%
MFCD00011530
CN(C)CC1=CC(=CC=C1)OC
165.236
165.24

Chemical Identifiers

15184-99-3
165.236
IENUHHADGQLQLU-UHFFFAOYSA-N
C10H15NO
MFCD00011530
547434
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents