missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5497003

 View more versions of this product

Catalog No. AAH5497003



Description

Description

3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Specifications

Specifications

3-Methoxy-N-methylbenzylamine
1.014
100°C (212°F)
1.529
1 g
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine
FIFKRPFWLHBMHL-UHFFFAOYSA-O
1-(3-methoxyphenyl)-N-methylmethanamine
5152225
97%
41789-95-1
130°C to 133°C
C9H14NO
MFCD04618244
Air Sensitive
Soluble in chloroform,dichloromethane,ethyl acetate and methanol.
C[NH2+]CC1=CC=CC(OC)=C1
152.22
151.21

Chemical Identifiers

41789-95-1
152.22
FIFKRPFWLHBMHL-UHFFFAOYSA-O
5152225
C[NH2+]CC1=CC=CC(OC)=C1
C9H14NO
MFCD04618244
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine
1-(3-methoxyphenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P270-P280-P285-P301+P312-P304+P341-P305+P351+P338-P310-P330-P342+P311-P501c

H302-H318-H334

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only