missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5497006

 View more versions of this product

Catalog No. AAH5497006



Description

Description

Applications
3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Specifications

Specifications

3-Methoxy-N-methylbenzylamine
100°C (212°F)
Air Sensitive
5g
C9H13NO
1-3-methoxyphenyl-n-methyl-methanamine
FIFKRPFWLHBMHL-UHFFFAOYSA-N
1-(3-methoxyphenyl)-N-methylmethanamine
5152225
97%
1.014
1.529
130°C to 133°C
41789-95-1
MFCD04618244
Soluble in chloroform,dichloromethane,ethyl acetate and methanol.
CNCC1=CC(=CC=C1)OC
151.209
151.21

Chemical Identifiers

41789-95-1
151.209
FIFKRPFWLHBMHL-UHFFFAOYSA-N
5152225
CNCC1=CC(=CC=C1)OC
C9H13NO
MFCD04618244
1-3-methoxyphenyl-n-methyl-methanamine
1-(3-methoxyphenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P270-P280-P285-P301+P312-P304+P341-P305+P351+P338-P310-P330-P342+P311-P501c

H302-H318-H334

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents