Please login to your online account to display your discounted pricing

3-methoxy-n-methylaniline 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC382230010

 View more versions of this product

Catalog No. AC382230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 137.18
Quantity 1g
Color Brown
Density 1.0490g/mL
CAS Min % 96.0
CAS Max % 100.0
Synonym N-Methyl-m-anisidine
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H11NO
Chemical Name or Material 3-Methoxy-N-methylaniline
Assay Percent Range 97%
MDL Number MFCD05665953
CAS 14318-66-2
Storage Information Product may darken on storage
Packaging Glass bottle
Refractive Index 1.5680 to 1.5700 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula CH3OC6H4NHCH3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

Specifications

Chemical Name or Material 3-Methoxy-N-methylaniline
CAS 14318-66-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H11NO
Linear Formula CH3OC6H4NHCH3
Formula Weight 137.18
MDL Number MFCD05665953
Synonym N-Methyl-m-anisidine
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

SDS & Safety Documents