Please login to your online account to display your discounted pricing

3-methoxy-n-methylaniline 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methoxy-N-methylaniline
CAS 14318-66-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H11NO
Linear Formula CH3OC6H4NHCH3
Formula Weight 137.18
MDL Number MFCD05665953
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382230010

  SDS
fisher scientific acros
AC382230010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 137.18
Quantity 1g
Color Brown
Density 1.0490g/mL
CAS Min % 96.0
CAS Max % 100.0
Synonym N-Methyl-m-anisidine
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Methoxy-N-methylaniline
Storage Information Product may darken on storage
Assay Percent Range 97%
Molecular Formula C8H11NO
MDL Number MFCD05665953
CAS 14318-66-2
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3OC6H4NHCH3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
Refractive Index 1.5680 to 1.5700 (20°C, 589nm)
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.