Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)phenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenylacetonitrile
Molecular Formula C10H8F3NO
Formula Weight 215.18
MDL Number MFCD09025415
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31621MD

  SDS
alfa aesar™
H31621MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3162103

  SDS
alfa aesar™
H3162103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.18
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenylacetonitrile
Assay 97%
Molecular Formula C10H8F3NO
MDL Number MFCD09025415
UN Number UN3276
Quantity 250mg
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.