missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)phenylacetic acid, 97%

$155.58 - $517.59

Chemical Identifiers

CAS 916421-04-0
Molecular Formula C10H9F3O3
Formula Weight 234.18
MDL Number MFCD09025416
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3198503 SDS Alfa Aesar™
H3198503
1g Each for $517.59
AAH31985MD SDS Alfa Aesar™
H31985MD
250mg Each for $155.58
Specifications Description & Specifications

Chemical Identifiers

916421-04-0
234.18
3-Methoxy-5-(trifluoromethyl)phenylacetic acid
C10H9F3O3
MFCD09025416

Specifications

916421-04-0
234.18
73° to 75°C
3-Methoxy-5-(trifluoromethyl)phenylacetic acid
C10H9F3O3
MFCD09025416
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning