Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)cinnamic acid, 97%

$89.51 - $299.85

Chemical Identifiers

Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)cinnamic acid
Molecular Formula C11H9F3O3
Formula Weight 246.18
MDL Number MFCD11519315
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3213303

  SDS
alfa aesar™
H3213303
1g Each for $299.85

AAH32133MD

  SDS
alfa aesar™
H32133MD
250mg Each for $89.51
Description & Specifications

Specifications

Formula Weight 246.18
Molecular Formula C11H9F3O3
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)cinnamic acid
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD11519315
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 155° to 157°C

Applications
3-Methoxy-5-(trifluoromethyl)cinnamic acid, is used as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.