Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)benzyl alcohol, 97%

Chemical Identifiers

Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzyl alcohol
Molecular Formula C9H9F3O2
Formula Weight 206.16
MDL Number MFCD09025413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3255503

  SDS
alfa aesar™
H3255503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3255506

  SDS
alfa aesar™
H3255506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.16
MDL Number MFCD09025413
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzyl alcohol
Health Hazard 1 Warning
Molecular Formula C9H9F3O2
Assay 97%
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 32° to 35°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.