Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)benzoic acid, 97%

$114.51 - $461.07

Chemical Identifiers

Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzoic acid
CAS 53985-48-1
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD09025409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3243003

  SDS
alfa aesar™
H3243003
1g Each for $114.51

AAH3243006

  SDS
alfa aesar™
H3243006
5g Each for $461.07
Description & Specifications

Specifications

Formula Weight 220.15
Quantity 1g
MDL Number MFCD09025409
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H7F3O3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 141° to 143°C
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzoic acid
Health Hazard 1 Warning
Assay 97%
CAS 53985-48-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.