Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-5-(trifluoromethyl)benzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzaldehyde
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD08458144
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3279106

  SDS
alfa aesar™
H3279106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.15
MDL Number MFCD08458144
Quantity 5g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Assay 97%
Molecular Formula C9H7F3O2
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.