missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-4-methylbenzonitrile, TCI America™

$55.00 - $100.83

Chemical Identifiers

CAS 3556-60-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD03428590
InChI Key QLJZMAGXHHXXMS-UHFFFAOYSA-N
Synonym 4-Cyano-2-methoxytoluene, 5-Cyano-2-methylanisole
PubChem CID 14752994
IUPAC Name 3-methoxy-4-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)C#N)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19771G View Documents TCI America
M19771G
1g Each for $55.00
 
M19775G View Documents TCI America
M19775G
5g Each for $100.83
 
Specifications

Chemical Identifiers

3556-60-3
147.177
QLJZMAGXHHXXMS-UHFFFAOYSA-N
14752994
CC1=C(C=C(C=C1)C#N)OC
C9H9NO
MFCD03428590
4-Cyano-2-methoxytoluene, 5-Cyano-2-methylanisole
3-methoxy-4-methylbenzonitrile

Specifications

White-Yellow
1g
C9H9NO
3439
QLJZMAGXHHXXMS-UHFFFAOYSA-N
3-methoxy-4-methylbenzonitrile
14752994
Crystal-Powder at 20°C
3-Methoxy-4-methylbenzonitrile
54°C
3556-60-3
MFCD03428590
4-Cyano-2-methoxytoluene, 5-Cyano-2-methylanisole
CC1=C(C=C(C=C1)C#N)OC
147.177
147.18
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No