missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-4-methylbenzeneboronic acid, 97%

$67.31 - $189.99

Chemical Identifiers

CAS 917757-15-4
Molecular Formula C8H11BO3
Formula Weight 165.99
MDL Number MFCD08274479
Synonym 3-Methoxy-4-methylphenylboronic acid; 3-Methoxy-p-tolylboronic acid
Chemical Name or Material 3-Methoxy-4-methylbenzeneboronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2875306 SDS Alfa Aesar™
H2875306
5g Each for $189.99
AAH2875303 SDS Alfa Aesar™
H2875303
1g Each for $67.31
Specifications

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

917757-15-4
165.99
3-Methoxy-4-methylphenylboronic acid; 3-Methoxy-p-tolylboronic acid
C8H11BO3
MFCD08274479
3-Methoxy-4-methylbenzeneboronic acid

Specifications

192° to 194°C
917757-15-4
165.99
3-Methoxy-4-methylphenylboronic acid; 3-Methoxy-p-tolylboronic acid
97%
5g
C8H11BO3
MFCD08274479
3-Methoxy-4-methylbenzeneboronic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning