missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 603122-41-4
Molecular Formula C9H11BO5
Molecular Weight (g/mol) 210
InChI Key YCXPWNGIPLGCOJ-UHFFFAOYSA-N
Synonym 3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, methyl 4-borono-2-methoxybenzoate, ycxpwngiplgcoj-uhfffaoysa-n
PubChem CID 17750256
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445480010
SDS View Documents
Thermo Fisher Scientific
AC445480010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445480050
SDS View Documents
Thermo Fisher Scientific
AC445480050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

603122-41-4
210
3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, methyl 4-borono-2-methoxybenzoate, ycxpwngiplgcoj-uhfffaoysa-n
C9H11BO5
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
17750256

Specifications

Glass Bottle
603122-41-4
C9H11BO5
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
17750256
97%
1g
96% min. (HPLC)
3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, methyl 4-borono-2-methoxybenzoate, ycxpwngiplgcoj-uhfffaoysa-n
210
210
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS