missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445480010

 View more versions of this product

Catalog No. AC445480010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
1g
96% min. (HPLC)
3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, methyl 4-borono-2-methoxybenzoate, ycxpwngiplgcoj-uhfffaoysa-n
210
210
Glass Bottle
603122-41-4
C9H11BO5
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
17750256
97%

Chemical Identifiers

603122-41-4
210
3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, methyl 4-borono-2-methoxybenzoate, ycxpwngiplgcoj-uhfffaoysa-n
C9H11BO5
YCXPWNGIPLGCOJ-UHFFFAOYSA-N
17750256
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents