Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-4-(4-piperidinyloxy)benzonitrile, 97%

$182.11 - $182.11

Chemical Identifiers

Chemical Name or Material 3-Methoxy-4-(4-piperidinyloxy)benzonitrile
CAS 902837-12-1
Molecular Formula C13H16N2O2
Formula Weight 232.28g/mol
MDL Number MFCD08061089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5096003

 
alfa aesar™
H50960-03
Each for $182.11
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 3-Methoxy-4-(4-piperidinyloxy)benzonitrile
CAS 902837-12-1
Assay 97%
Molecular Formula C13H16N2O2
Formula Weight 232.28g/mol
MDL Number MFCD08061089
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.