missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-2-nitrotoluene 98.0+%, TCI America™

$97.11 - $288.03

Chemical Identifiers

CAS 5345-42-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007179
InChI Key MGBRGNWARSQECY-UHFFFAOYSA-N
Synonym #, 2-nitro-3-methylanisole, 3-methoxy-1-methyl-2-nitrobenzene, 3-methoxy-2-nitrotoluene, 3-methyl-2-nitroanisole, acmc-209l7h, anisole, 3-methyl-2-nitro, benzene, 1-methoxy-3-methyl-2-nitro, pubchem12903
PubChem CID 79291
IUPAC Name 1-methoxy-3-methyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)OC)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11965G View Documents Tci America
M11965G
5g Each for $97.11
 
M119625G View Documents Tci America
M119625G
25g Each for $288.03
 
Specifications

Chemical Identifiers

5345-42-6
167.164
MGBRGNWARSQECY-UHFFFAOYSA-N
79291
CC1=C(C(=CC=C1)OC)[N+](=O)[O-]
C8H9NO3
MFCD00007179
#, 2-nitro-3-methylanisole, 3-methoxy-1-methyl-2-nitrobenzene, 3-methoxy-2-nitrotoluene, 3-methyl-2-nitroanisole, acmc-209l7h, anisole, 3-methyl-2-nitro, benzene, 1-methoxy-3-methyl-2-nitro, pubchem12903
1-methoxy-3-methyl-2-nitrobenzene

Specifications

170°C
49°C
5345-42-6
MFCD00007179
MGBRGNWARSQECY-UHFFFAOYSA-N
1-methoxy-3-methyl-2-nitrobenzene
79291
Crystal-Powder at 20°C
3-Methoxy-2-nitrotoluene
Yellow
5g
C8H9NO3
#, 2-nitro-3-methylanisole, 3-methoxy-1-methyl-2-nitrobenzene, 3-methoxy-2-nitrotoluene, 3-methyl-2-nitroanisole, acmc-209l7h, anisole, 3-methyl-2-nitro, benzene, 1-methoxy-3-methyl-2-nitro, pubchem12903
CC1=C(C(=CC=C1)OC)[N+](=O)[O-]
167.164
167.16
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0799

TSCA : No