missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-2-nitrobenzoic Acid 99.0+%, TCI America™

Manufacturer:  TCI America M103610G

 View more versions of this product

Catalog No. M103610G



Specifications

Specifications

3-Methoxy-2-nitrobenzoic Acid
10g
C8H7NO5
2-nitro-3-methoxybenzoic acid
COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O
78632
Crystal-Powder at 20°C
White
4920-80-3
MFCD00007143
YMOMYSDAOXOCID-UHFFFAOYSA-N
197.146
197.15
≥99.0% (T)

Chemical Identifiers

4920-80-3
197.146
YMOMYSDAOXOCID-UHFFFAOYSA-N
78632
C8H7NO5
MFCD00007143
2-nitro-3-methoxybenzoic acid
COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents