Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-2-nitrobenzoic acid, 99%

$40.99 - $112.39

Chemical Identifiers

Chemical Name or Material 3-Methoxy-2-nitrobenzoic acid
CAS 4920-80-3
Molecular Formula C8H7NO5
Formula Weight 197.15
MDL Number MFCD00007143
Beilstein 2214986
Synonym 2-Nitro-m-anisic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0353103

  SDS
alfa aesar™
L0353103
1g Each for $40.99

AAL0353106

  SDS
alfa aesar™
L0353106
5g Each for $112.39
Description & Specifications

Specifications

Formula Weight 197.15
Chemical Name or Material 3-Methoxy-2-nitrobenzoic acid
Molecular Formula C8H7NO5
MDL Number MFCD00007143
CAS 4920-80-3
Synonym 2-Nitro-m-anisic acid
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Beilstein 2214986
Melting Point 255° to 257°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.