Please login to your online account to display your discounted pricing

3-Methoxy-2,4,5-trifluorobenzoic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methoxy-2, 4, 5-trifluorobenzoic acid
Name Note 97%
CAS 11281-65-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5F3O3
Linear Formula CH3OC6H(F)3COOH
Formula Weight 206.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC345060050

  SDS
thermo fisher scientific
AC345060050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 206.12
Quantity 5g
Color White
TSCA TSCA
Melting Point 114° to 119°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass Bottle
Chemical Name or Material 3-Methoxy-2, 4, 5-trifluorobenzoic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H5F3O3
Linear Formula CH3OC6H(F)3COOH
MDL Number MFCD00153201
CAS 11281-65-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.