missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-2,4,5-trifluorobenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC345060050

 View more versions of this product

Catalog No. AC345060050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Methoxy-2, 4, 5-trifluorobenzoic acid
11281-65-5
100.0
Authentic
≥96.0%
C8H5F3O3
206.12
White
5g
97%
96.0
Glass Bottle
Powder
97%
CH3OC6H(F)3COOH
MFCD00153201
114° to 119°C

Chemical Identifiers

11281-65-5
100.0
C8H5F3O3
206.12
3-Methoxy-2, 4, 5-trifluorobenzoic acid, 97%
96.0
97%
CH3OC6H(F)3COOH
MFCD00153201
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA