missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-2(1H)-pyridone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163540010

 View more versions of this product

Catalog No. AC163540010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Methoxy-2(1H)-pyridone
Glass bottle
113.0°C to 115.0°C
20928-63-6
100.0
C6H7NO2
LKIMDXQLHFCXQF-UHFFFAOYSA-N
3-methoxy-1H-pyridin-2-one
88731
Powder or Crystalline Powder
Authentic
White to Brown
1g
96.0
96% min. (HPLC)
2 1h-pyridinone, 3-methoxy
COC1=CC=CNC1=O
125.13
125.13
97%

Chemical Identifiers

20928-63-6
125.13
2 1h-pyridinone, 3-methoxy
3-methoxy-1H-pyridin-2-one
C6H7NO2
LKIMDXQLHFCXQF-UHFFFAOYSA-N
88731
COC1=CC=CNC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-116-1

SDS Documents