Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-2(1H)-pyridone, 97%

$103.14 - $103.14

Chemical Identifiers

Chemical Name or Material 3-Methoxy-2(1H)-pyridone
CAS 20928-63-6
Molecular Formula C6H7NO2
Formula Weight 125.13
MDL Number MFCD00006015
Beilstein 1447118
Synonym 2-Hydroxy-3-methoxypyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0708003

 
alfa aesar™
L0708003
1g Each for $103.14
Description & Specifications

Specifications

Formula Weight 125.13
Chemical Name or Material 3-Methoxy-2(1H)-pyridone
Assay 97%
MDL Number MFCD00006015
CAS 20928-63-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H7NO2
Beilstein 1447118
Synonym 2-Hydroxy-3-methoxypyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 112° to 118°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.