missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxy-1-propanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 137150500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Methoxy-1-propanol | |
97.5 | |
0.9420g/mL | |
38°C | |
97.5% min. (GC) | |
C4H10O2 | |
CH3O(CH2)3OH | |
0.942 | |
JDFDHBSESGTDAL-UHFFFAOYSA-N | |
3-methoxypropan-1-ol | |
74116 | |
98% |
1589-49-7 | |
100.0 | |
149.0°C to 150.0°C | |
Authentic | |
Glass bottle | |
1.4120 to 1.4140 (20°C, 589nm) | |
50g | |
3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol | |
COCCCO | |
90.12 | |
90.12 |
Chemical Identifiers
1589-49-7 | |
90.12 | |
3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol | |
3-methoxypropan-1-ol |
C4H10O2 | |
JDFDHBSESGTDAL-UHFFFAOYSA-N | |
74116 | |
COCCCO |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
RUO â Research Use Only