missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxy-1-butanol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125791000
Specifications
3-Methoxy-1-butanol | |
46°C | |
Glass bottle | |
0.92 | |
161.0°C | |
-85.0°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
CH3CH(OCH3)CH2CH2OH | |
1987 | |
Solubility in water: soluble. Other solubilities: miscible with most common organic solvents | |
CC(CCO)OC | |
104.15 | |
104.15 | |
99% |
0.9200g/mL | |
Authentic | |
1.4150 to 1.4165 | |
0.2% max. | |
Colorless | |
100mL | |
2517-43-3 | |
100.0 | |
C5H12O2 | |
MFCD00002931 | |
1-butanol, 3-methoxy, 1-butanol, 3-methoxy-, r, 1-butanol, 3-methoxy-,-, 3-methoxy-1-butanol, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutanol, 3-methoxybutyl alcohol, ccris 8976, methoxybutanol | |
JSGVZVOGOQILFM-UHFFFAOYSA-N | |
3-methoxybutan-1-ol | |
17291 | |
Liquid |
Chemical Identifiers
2517-43-3 | |
104.15 | |
JSGVZVOGOQILFM-UHFFFAOYSA-N | |
17291 | |
CC(CCO)OC |
C5H12O2 | |
MFCD00002931 | |
1-butanol, 3-methoxy, 1-butanol, 3-methoxy-, r, 1-butanol, 3-methoxy-,-, 3-methoxy-1-butanol, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutanol, 3-methoxybutyl alcohol, ccris 8976, methoxybutanol | |
3-methoxybutan-1-ol |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
GHS Signal Word: Warning
EINECSNumber : 219-741-8
TSCA : TSCA