missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-1,2-propanediol 98.0+%, TCI America™

Manufacturer:  TCI America M0125500ML

 View more versions of this product

Catalog No. M0125500ML



Specifications

Specifications

3-Methoxy-1,2-propanediol
Yellow
623-39-2
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
3-methoxypropane-1,2-diol
92822
Clear Liquid at 20°C
220°C
500mL
C4H10O3
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
COCC(CO)O
106.121
106.12
≥98.0% (GC)

Chemical Identifiers

623-39-2
106.121
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
92822
COCC(CO)O
C4H10O3
MFCD00004719
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
3-methoxypropane-1,2-diol
Safety and Handling

Safety and Handling

RTECSNumber : TY8170000

TSCA : Yes

Documents