missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxy-1,2-propanediol, TCI America™

$125.28 - $1,066.41

Chemical Identifiers

CAS 623-39-2
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.121
MDL Number MFCD00004719
InChI Key PSJBSUHYCGQTHZ-UHFFFAOYSA-N
Synonym 3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
PubChem CID 92822
IUPAC Name 3-methoxypropane-1,2-diol
SMILES COCC(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M012525ML View Documents TCI America
M012525ML
25mL Each for $125.28
 
M0125500ML View Documents TCI America
M0125500ML
500mL Each for $1,066.41
 
Specifications

Chemical Identifiers

623-39-2
106.121
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
92822
COCC(CO)O
C4H10O3
MFCD00004719
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
3-methoxypropane-1,2-diol

Specifications

220°C
25mL
C4H10O3
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
COCC(CO)O
106.121
106.12
≥98.0% (GC)
Yellow
623-39-2
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
3-methoxypropane-1,2-diol
92822
Clear Liquid at 20°C
3-Methoxy-1,2-propanediol
Safety and Handling

Safety and Handling

RTECSNumber : TY8170000

TSCA : Yes