Please login to your online account to display your discounted pricing

3-Methoxy-1,2-propanediol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338390010

 View more versions of this product

Catalog No. AC338390010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 106.12
Quantity 1g
Color Clear colorless
Density 1.12g/mL
CAS Max % 100.0
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Molecular Formula C4H10O3
Linear Formula H3COCHCH(OH)CHOH
Chemical Name or Material 3-Methoxy-1, 2-propanediol
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00004719
CAS 623-39-2
CAS Min % 97.5
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Methoxy-1, 2-propanediol
CAS 623-39-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H10O3
Linear Formula H3COCHCH(OH)CHOH
Formula Weight 106.12
MDL Number MFCD00004719