Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxy-1,2-propanediol, 97%

$32.54 - $85.58

Chemical Identifiers

Chemical Name or Material 3-Methoxy-1,2-propanediol
CAS 623-39-2
Molecular Formula C4H10O3
Formula Weight 106.12
MDL Number MFCD00004719
Beilstein 1733242
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0801103

  SDS
alfa aesar™
L0801103
1g Each for $32.54

AAL0801106

  SDS
alfa aesar™
L0801106
5g Each for $85.58
Description & Specifications

Specifications

Formula Weight 106.12
Chemical Name or Material 3-Methoxy-1,2-propanediol
Molecular Formula C4H10O3
MDL Number MFCD00004719
Assay 97%
Beilstein 1733242
Quantity 1g
TSCA TSCA
Flash Point >110°C (230°F)
Boiling Point 218° to 220°C
CAS 623-39-2