missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride, TCI America™
$64.17 - $189.45
Chemical Identifiers
CAS | 41762-76-9 |
---|---|
Molecular Formula | C5H7ClN2O4S |
Molecular Weight (g/mol) | 226.631 |
MDL Number | MFCD00272360 |
InChI Key | ZWTPALHHEULAPI-UHFFFAOYSA-N |
Synonym | 1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone |
PubChem CID | 3016299 |
IUPAC Name | 3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride |
SMILES | CS(=O)(=O)N1CCN(C1=O)C(=O)Cl |
Chemical Identifiers
41762-76-9 | |
226.631 | |
ZWTPALHHEULAPI-UHFFFAOYSA-N | |
3016299 | |
CS(=O)(=O)N1CCN(C1=O)C(=O)Cl |
C5H7ClN2O4S | |
MFCD00272360 | |
1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone | |
3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride |
Specifications
White-Yellow | |
5g | |
C5H7ClN2O4S | |
3261 | |
ZWTPALHHEULAPI-UHFFFAOYSA-N | |
3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride | |
3016299 | |
Crystal-Powder at 20°C | |
3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride |
180°C | |
41762-76-9 | |
MFCD00272360 | |
1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone | |
CS(=O)(=O)N1CCN(C1=O)C(=O)Cl | |
226.631 | |
226.63 | |
≥98.0% (T) |
Safety and Handling
TSCA : No