Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Mercaptopropyl)methyldimethoxysilane, 95%

$26.16 - $862.00

Chemical Identifiers

Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
CAS 31001-77-1
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
Formula Weight 180.34
MDL Number MFCD00053923
Synonym 3-(Dimethoxymethylsilyl)-1-propanethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42426AB

 
alfa aesar™
42426AB
10mL Each for $26.16

AA42426K2

 
alfa aesar™
42426K2
1L Each for $862.00

AA42426AD

 
alfa aesar™
42426AD
50mL Each for $74.51

AA42426AK

 
alfa aesar™
42426AK
250mL Each for $275.53
Description & Specifications

Specifications

Formula Weight 180.34
Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
TSCA TSCA
Physical Form Liquid
Refractive Index 1.4500
Assay 95%
Sensitivity Moisture Sensitive
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
MDL Number MFCD00053923
CAS 31001-77-1
Synonym 3-(Dimethoxymethylsilyl)-1-propanethiol
Boiling Point 96°C (30mmHg)
Color Colorless
Quantity 10mL
Flash Point 93°C (199°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Density 1.000g/mL
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.