missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Mercaptoisobutyric Acid, TCI America™

$197.09 - $592.80

Chemical Identifiers

CAS 26473-47-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00671555
InChI Key MHRDCHHESNJQIS-UHFFFAOYSA-N
Synonym 3-Mercapto-2-methylpropionic Acid
PubChem CID 152910
IUPAC Name 2-methyl-3-sulfanylpropanoic acid
SMILES CC(CS)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12005G View Documents TCI America
M12005G
5g Each for $197.09
 
M120025G View Documents TCI America
M120025G
25g Each for $592.80
 
Specifications

Chemical Identifiers

26473-47-2
120.166
MHRDCHHESNJQIS-UHFFFAOYSA-N
152910
CC(CS)C(=O)O
C4H8O2S
MFCD00671555
3-Mercapto-2-methylpropionic Acid
2-methyl-3-sulfanylpropanoic acid

Specifications

122°C
5g
C4H8O2S
3-Mercapto-2-methylpropionic Acid
CC(CS)C(=O)O
120.166
120.17
≥98.0% (GC,T)
Yellow
26473-47-2
MFCD00671555
MHRDCHHESNJQIS-UHFFFAOYSA-N
2-methyl-3-sulfanylpropanoic acid
152910
Clear Liquid at 20°C
3-Mercaptoisobutyric Acid
Safety and Handling

Safety and Handling

TSCA : No